Assay Details
Target Gene Details
Entrez Gene ID: | 244209 |
Gene Name: | cytochrome P450, family 2, subfamily r, polypeptide 1 |
Gene Aliases: |
- |
Location: |
Chr.7:114550123-114565618 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Cyp2r1 | NM_177382.4 | 3 | 699 | NP_796356.2 |
NR_110407.1 | ||||
NR_110408.1 | ||||
AK050453.1 | BAC34265.1 | |||
AY323818.1 | 3 | 699 | AAQ23115.1 | |
BC108961.1 | 3 | 699 | AAI08962.1 | |
BC108962.1 | AAI08963.1 |
More Information
Set Membership: |
Intragenic Exonic |