Assay Details
Target Gene Details
Entrez Gene ID: | 26433 |
Gene Name: | procollagen-lysine, 2-oxoglutarate 5-dioxygenase 3 |
Gene Aliases: |
AI414586, LH3 |
Location: |
Chr.5:136987019-136996646 on Build GRCm38 |
Assay Gene Location: | Within Exon 19 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Plod3 | NM_011962.3 | 19 | 2951 | NP_036092.1 |
AF046783.1 | 19 | 2359 | AAD54618.1 | |
AK013195.1 | 19 | 2497 | BAB28704.1 | |
AK033360.1 | 19 | 2496 | BAC28246.1 | |
AK088948.1 | 19 | 2368 | BAC40669.1 | |
AK193849.1 | 1 | 100 | ||
AK194341.1 | 1 | 100 | ||
AK197520.1 | 1 | 100 | ||
AK199802.1 | 1 | 100 | ||
AK200122.1 | 1 | 100 | ||
AK201573.1 | 1 | 100 | ||
AK206950.1 | 1 | 100 | ||
AK209058.1 | 1 | 100 | ||
AK215272.1 | 1 | 339 | ||
AK215677.1 | 1 | 100 | ||
AK217108.1 | 1 | 100 | ||
AK219112.1 | 1 | 100 | ||
AK220112.1 | 1 | 100 | ||
BB692664.1 | 1 | 212 | ||
BC043047.1 | 19 | 2950 | AAH43047.1 | |
BC054734.1 | 19 | 2951 | AAH54734.1 |
More Information
Set Membership: |
Intragenic Exonic |