Assay Details
Target Gene Details
Entrez Gene ID: | 72898 |
Gene Name: | aspartate beta-hydroxylase domain containing 2 |
Gene Aliases: |
2900006N09Rik, 9230106G13Rik |
Location: |
Chr.5:112384354-112395379 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Asphd2 | NM_028386.1 | 1 | 704 | NP_082662.1 |
BC046606.1 | 1 | 704 | AAH46606.1 |
More Information
Set Membership: |
Intragenic Exonic |