Assay Details
Target Gene Details
Entrez Gene ID: | 14748 |
Gene Name: | G-protein coupled receptor 3 |
Gene Aliases: |
Gpcr20, Gpcr21, Gpcr3 |
Location: |
Chr.4:133209340-133212536 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Gpr3 | NM_008154.3 | 2 | 1135 | NP_032180.1 |
AK140302.1 | 2 | 1106 | BAE24325.1 | |
AK140464.1 | 2 | 1089 | BAE24399.1 | |
AK158917.1 | 2 | 1098 | BAE34727.1 | |
BC138248.1 | 1 | 1457 | ||
BC138250.1 | 1 | 1457 | ||
D21062.1 | 1 | 1510 | BAA04641.1 |
More Information
Set Membership: |
Intragenic Exonic |