Assay Details
Target Gene Details
Entrez Gene ID: | 65973 |
Gene Name: | aspartate-beta-hydroxylase |
Gene Aliases: |
2310005F16Rik, 3110001L23Rik, AI848629, AW261690, AW561901, BAH, C79816, cI-37 |
Location: |
Chr.4:9449085-9669344 on Build GRCm38 |
Assay Gene Location: | Within Exon 30 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Asph | NM_001177849.1 | 23 | 5182 | NP_001171320.1 |
NM_001290367.1 | 23 | 4989 | NP_001277296.1 | |
NM_023066.3 | 24 | 5230 | NP_075553.2 | |
AF289487.1 | 24 | 5146 | AAG40809.1 |
Target Gene Details
Entrez Gene ID: | 74438 |
Gene Name: | clavesin 1 |
Gene Aliases: |
4933402J24Rik, Clvl1, Rlbp1l1 |
Location: |
Chr.4:9144756-9451691 on Build GRCm38 |
Assay Gene Location: | Within Exon 6 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Clvs1 | NM_028940.2 | 6 | 2456 | NP_083216.1 |
AK032549.1 | 6 | 2446 | ||
AK136083.1 | 5 | 1991 | BAE22811.1 | |
AK147767.1 | 1 | 1058 | ||
BC062923.1 | 6 | 2411 | AAH62923.1 | |
BC100556.1 | 4 | 1676 | AAI00557.1 |
More Information
Set Membership: |
Intragenic Exonic |