Assay Details
Target Gene Details
Entrez Gene ID: | 65973 |
Gene Name: | aspartate-beta-hydroxylase |
Gene Aliases: |
2310005F16Rik, 3110001L23Rik, AI848629, AW261690, AW561901, BAH, C79816, cI-37 |
Location: |
Chr.4:9449085-9669344 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Asph | NM_001177849.1 | 1 | 153 | NP_001171320.1 |
NM_001177850.1 | 1 | 153 | NP_001171321.1 | |
NM_001177851.1 | 1 | 153 | NP_001171322.1 | |
NM_001177855.1 | 1 | 153 | NP_001171326.1 | |
NM_001177856.1 | 1 | 153 | NP_001171327.1 | |
NM_001290367.1 | NP_001277296.1 | |||
NM_023066.3 | 1 | 153 | NP_075553.2 | |
AF289486.1 | 1 | 75 | AAG40808.1 | |
AF289487.1 | 1 | 75 | AAG40809.1 | |
AF289488.1 | 1 | 75 | AAG40810.1 | |
AK030293.1 | 1 | 77 | BAC26882.1 | |
AK032420.1 | 1 | 76 | ||
AK035735.1 | BAC29171.1 | |||
AK049506.1 | 1 | 77 | BAC33783.1 | |
AK139566.1 | 1 | 77 | ||
AK151787.1 | 1 | 76 | BAE30688.1 | |
AK160984.1 | 1 | 78 | BAE36131.1 | |
AK165260.1 | 1 | 99 | BAE38110.1 | |
BC015281.1 | 1 | 62 | AAH15281.1 | |
BC061098.1 | 1 | 99 | AAH61098.1 | |
BY268464.1 | 1 | 154 |
More Information
Set Membership: |
Intragenic Exonic |