Assay Details
Target Gene Details
Entrez Gene ID: | 19275 |
Gene Name: | protein tyrosine phosphatase, receptor type, N |
Gene Aliases: |
IA-2, mIA-A, mKIAA4064 |
Location: |
Chr.1:75247035-75264497 on Build GRCm38 |
Assay Gene Location: | Within Exon 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Ptprn | NM_008985.2 | 5 | 687 | NP_033011.2 |
AF332083.1 | 2 | 291 | AAK56111.1 | |
AF332084.1 | 2 | 291 | AAK56112.1 | |
AK041296.1 | 5 | 693 | ||
AK076223.1 | 5 | 666 | ||
AK139398.1 | 5 | 687 | BAE23994.1 | |
AK163141.1 | 5 | 663 | ||
AK220467.1 | 5 | 645 | BAD90494.1 | |
BC068165.1 | 5 | 619 | AAH68165.1 | |
BM950464.1 | 3 | 353 | ||
DQ832283.1 | 5 | 664 | ||
U11812.1 | 5 | 611 | AAA52102.1 | |
X74438.1 | 5 | 704 | CAA52453.1 |
More Information
Set Membership: |
Intragenic Exonic |