Assay Details
Target Gene Details
Entrez Gene ID: | 75060 |
Gene Name: | RIKEN cDNA 4930506C21 gene |
Gene Aliases: |
- |
Location: |
Chr.17:8293366-8311118 on Build GRCm38 |
Assay Gene Location: | Overlaps - Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
4930506C21Rik | NR_073374.1 | |||
AK043779.1 |
Target Gene Details
Entrez Gene ID: | 106489 |
Gene Name: | SFT2 domain containing 1 |
Gene Aliases: |
5630401J11Rik, AA409698 |
Location: |
Chr.17:8311103-8327442 on Build GRCm38 |
Assay Gene Location: | Overlaps - Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Sft2d1 | NM_134114.2 | NP_598875.2 | ||
BC091770.1 | AAH91770.1 |