Assay Details
Target Gene Details
Entrez Gene ID: | 14229 |
Gene Name: | FK506 binding protein 5 |
Gene Aliases: |
D17Ertd592e, Dit1, FKBP-5, FKBP51 |
Location: |
Chr.17:28398753-28486149 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 7 - Exon 8 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Fkbp5 | NM_010220.4 | NP_034350.1 | ||
AK169795.1 | BAE41371.1 | |||
BC015260.1 | AAH15260.1 | |||
BU613862.1 | ||||
CF949740.1 | ||||
CT010372.1 | CAJ18579.1 | |||
U16959.1 | AAA86983.1 | |||
U36220.1 | AAA89162.1 |
More Information
Set Membership: |
Intragenic Non-exonic |