Assay Details
Target Gene Details
Entrez Gene ID: | 13358 |
Gene Name: | solute carrier family 25 (mitochondrial carrier, citrate transporter), member 1 |
Gene Aliases: |
1300019P08Rik, 2610100G11Rik, AI194714, Ctp, Dgsj, Slc20a3 |
Location: |
Chr.16:17925211-17928219 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 8 - Exon 8 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Slc25a1 | NM_153150.2 | NP_694790.1 | ||
AK005070.1 | ||||
AK159289.1 | BAE34965.1 | |||
AK170191.1 | BAE41625.1 | |||
BC037087.1 | AAH37087.1 |
More Information
Set Membership: |
Intragenic Non-exonic |