Assay Details
Target Gene Details
Entrez Gene ID: | 81799 |
Gene Name: | C1q and tumor necrosis factor related protein 3 |
Gene Aliases: |
2310005P21Rik, CORS-26, CORS26, CTRP3, Corcs, Cors |
Location: |
Chr.15:10952332-10980162 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 5 - Intron 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
C1qtnf3 | NM_001204134.1 | NP_001191063.1 | ||
NM_030888.4 | NP_112150.1 | |||
AF246265.1 | AAG33704.1 | |||
AK029352.1 | BAC26412.1 | |||
AK075867.1 | BAC36016.1 | |||
AK081704.1 | BAC38301.1 | |||
AK128898.1 | ||||
BC055047.1 | AAH55047.1 | |||
BC055859.1 | AAH55859.1 | |||
DQ002396.1 | AAY21928.1 |