Assay Details
Target Gene Details
Entrez Gene ID: | 67285 |
Gene Name: | CWC27 spliceosome-associated protein homolog (S. cerevisiae) |
Gene Aliases: |
3110009E13Rik, NY-CO-10, Sdccag10 |
Location: |
Chr.13:104631322-104816964 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 11 - Exon 12 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Cwc27 | NM_026072.1 | NP_080348.1 | ||
AK014025.1 | BAB29120.1 | |||
AK075902.1 | BAC36042.1 | |||
AK077779.1 | BAC37003.1 | |||
AK166770.1 | BAE39007.1 | |||
BC025437.1 | AAH25437.1 |
More Information
Set Membership: |
Intragenic Non-exonic |