Assay Details
Target Gene Details
Entrez Gene ID: | 12763 |
Gene Name: | cytidine monophospho-N-acetylneuraminic acid hydroxylase |
Gene Aliases: |
- |
Location: |
Chr.13:24326980-24477289 on Build GRCm38 |
Assay Gene Location: | Within Exon 9 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Cmah | NM_001111110.2 | 7 | 1023 | NP_001104580.1 |
NM_001284519.1 | 8 | 1185 | NP_001271448.1 | |
NM_001284520.1 | 6 | 678 | NP_001271449.1 | |
NM_007717.5 | 6 | 678 | NP_031743.3 | |
AB061276.1 | 7 | 1007 | BAB91361.1 | |
AB061277.1 | 8 | 1169 | BAB91362.1 | |
AK040274.1 | 5 | 628 | ||
AK083394.1 | ||||
AK145110.1 | 5 | 826 | ||
AK145199.1 | 6 | 679 | BAE26290.1 | |
AK166486.1 | 6 | 679 | BAE38803.1 | |
BC055079.1 | 5 | 675 | AAH55079.1 | |
D21826.1 | 5 | 650 | BAA04850.1 |
More Information
Set Membership: |
Intragenic Exonic |