Assay Details
Target Gene Details
Entrez Gene ID: | 12763 |
Gene Name: | cytidine monophospho-N-acetylneuraminic acid hydroxylase |
Gene Aliases: |
- |
Location: |
Chr.13:24326980-24477289 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 14 - Exon 15 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Cmah | NM_001111110.2 | NP_001104580.1 | ||
NM_001284519.1 | NP_001271448.1 | |||
NM_001284520.1 | NP_001271449.1 | |||
NM_007717.5 | NP_031743.3 | |||
AB061276.1 | BAB91361.1 | |||
AB061277.1 | BAB91362.1 | |||
AK083394.1 | ||||
AK145110.1 | ||||
AK166486.1 | BAE38803.1 | |||
BC055079.1 | AAH55079.1 | |||
D21826.1 | BAA04850.1 |
More Information
Set Membership: |
Intragenic Non-exonic |