Assay Details
Target Gene Details
Entrez Gene ID: | 27364 |
Gene Name: | serine racemase |
Gene Aliases: |
Srs |
Location: |
Chr.11:74905047-74925798 on Build GRCm38 |
Assay Gene Location: | Within Exon 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Srr | NM_001163311.1 | 4 | 540 | NP_001156783.1 |
NM_013761.4 | 5 | 766 | NP_038789.1 | |
AF148321.1 | 4 | 421 | AAF08701.1 | |
AK017235.1 | 5 | 646 | ||
AK028034.1 | 4 | 423 | BAC25712.1 | |
AK031687.1 | 5 | 638 | BAC27514.1 | |
AK043738.1 | 5 | 672 | BAC31637.1 | |
AK043819.1 | 4 | 446 | ||
AK079343.1 | ||||
AK080830.1 | 5 | 646 | ||
AK157122.1 | 4 | 691 | BAE33968.1 | |
AK170096.1 | 5 | 652 | BAE41561.1 | |
AK170471.1 | 5 | 652 | ||
BC011164.1 | 5 | 643 | AAH11164.1 |
More Information
Set Membership: |
Intragenic Exonic |