Assay Details
Target Gene Details
Entrez Gene ID: | 66055 |
Gene Name: | splicing factor 3B, subunit 6 |
Gene Aliases: |
0610009D07Rik, 6030419K15Rik, AV001342, Sf3b14 |
Location: |
Chr.12:4817608-4827659 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Sf3b6 | NM_025323.2 | NP_079599.1 | ||
AK002375.1 | BAB22052.1 | |||
AK003683.1 | BAB22936.1 | |||
AK151648.1 | BAE30577.1 | |||
BC019535.1 | AAH19535.1 |
More Information
Set Membership: |
Intragenic Non-exonic |