Assay Details
Target Gene Details
Entrez Gene ID: | 104112 |
Gene Name: | ATP citrate lyase |
Gene Aliases: |
A730098H14Rik, AW538652 |
Location: |
Chr.11:100476352-100528000 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 28 - Exon 28 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Acly | NM_001199296.1 | NP_001186225.1 | ||
NM_134037.3 | NP_598798.1 | |||
AF332051.1 | AAK56080.1 | |||
AF332052.1 | AAK56081.1 | |||
AK043434.1 | ||||
AK132755.1 | BAE21336.1 | |||
AK135528.1 | ||||
AK136812.1 | BAE23135.1 | |||
AK169703.1 | BAE41315.1 | |||
BC005533.1 | AAH05533.1 | |||
BC021502.1 | AAH21502.1 | |||
BC056378.1 | AAH56378.1 | |||
BC065805.1 | AAH65805.1 |
More Information
Set Membership: |
Intragenic Non-exonic |