Assay Details
Target Gene Details
Entrez Gene ID: | 216343 |
Gene Name: | tryptophan hydroxylase 2 |
Gene Aliases: |
AU043594, Ntph |
Location: |
Chr.10:115078641-115185022 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 7 - Exon 7 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Tph2 | NM_173391.3 | NP_775567.2 | ||
AY090565.1 | AAM08923.1 | |||
BC120514.1 | ||||
BC125430.1 |