Assay Details
Target Gene Details
Entrez Gene ID: | 20397 |
Gene Name: | sphingosine phosphate lyase 1 |
Gene Aliases: |
AI428538, D10Xrf456, S1PL, Spl |
Location: |
Chr.10:61098642-61147686 on Build GRCm38 |
Assay Gene Location: | Within Exon 7 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Sgpl1 | NM_009163.3 | 7 | 743 | NP_033189.2 |
AF036894.1 | 6 | 613 | AAC03768.1 | |
AK036747.1 | 7 | 631 | BAC29562.1 | |
AK037789.1 | 7 | 628 | BAC29872.1 | |
AK043024.1 | 7 | 630 | BAC31437.1 | |
AK049342.1 | 7 | 651 | BAC33695.1 | |
AK150053.1 | 7 | 638 | BAE29269.1 | |
AK173137.1 | 7 | 672 | BAD32415.1 | |
BC026135.1 | 7 | 689 | AAH26135.1 | |
CT010214.1 | 6 | 553 | CAJ18422.1 |