Assay Details
Target Gene Details
Entrez Gene ID: | 216136 |
Gene Name: | ilvB (bacterial acetolactate synthase)-like |
Gene Aliases: |
5830463I21, AI415009 |
Location: |
Chr.10:78574345-78584502 on Build GRCm38 |
Assay Gene Location: | Within Exon 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Ilvbl | NM_173751.4 | 5 | 707 | NP_776112.1 |
AK030933.1 | 2 | 145 | ||
AK087956.1 | 4 | 581 | BAC40057.1 | |
AK133553.1 | 4 | 637 | BAE21720.1 | |
AK138031.1 | 4 | 556 | ||
AK143918.1 | 4 | 606 | BAE25603.1 | |
AK146075.1 | 4 | 635 | BAE26879.1 | |
AK169899.1 | 4 | 633 | ||
BC056459.1 | 5 | 665 | AAH56459.1 |
More Information
Set Membership: |
Intragenic Exonic |