Assay Details
Target Gene Details
Entrez Gene ID: | 13077 |
Gene Name: | cytochrome P450, family 1, subfamily a, polypeptide 2 |
Gene Aliases: |
CP12, Cyp1a1, P450-3 |
Location: |
Chr.9:57676937-57683655 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Cyp1a2 | NM_009993.3 | 4 | 1050 | NP_034123.1 |
BC018298.1 | 4 | 1030 | AAH18298.1 | |
BC054827.1 | 4 | 1043 | AAH54827.1 | |
K02589.1 | 4 | 1051 | AAA37509.1 | |
X00479.1 | 4 | 1051 | CAA25156.1 | |
X04283.1 | 3 | 1014 | CAA27832.1 |