Assay Details
Target Gene Details
Entrez Gene ID: | 13076 |
Gene Name: | cytochrome P450, family 1, subfamily a, polypeptide 1 |
Gene Aliases: |
AHH, AHRR, CP11, Cyp1a2, P450-1 |
Location: |
Chr.9:57687928-57703824 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Cyp1a1 | NM_001136059.2 | NP_001129531.1 | ||
NM_009992.4 | NP_034122.1 | |||
AK005000.1 | BAB23734.1 | |||
BC125440.1 | ||||
BC125444.1 | ||||
K02588.1 | AAA37506.1 | |||
Y00071.1 | CAA68277.1 |