Assay Details
Target Gene Details
Entrez Gene ID: | 23923 |
Gene Name: | aminoadipate aminotransferase |
Gene Aliases: |
AI875679, Aadt, KATII, Kat2, mKat-2 |
Location: |
Chr.8:60505870-60545677 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Aadat | NM_011834.2 | 2 | 199 | NP_035964.1 |
AF072376.1 | 2 | 199 | AAD39680.1 | |
AK075578.1 | 2 | 205 | BAC35833.1 | |
AK144042.1 | 2 | 152 | BAE25669.1 | |
BC012637.1 | 2 | 202 | AAH12637.1 |
More Information
Set Membership: |
Intragenic Exonic |