Assay Details
Target Gene Details
Entrez Gene ID: | 22228 |
Gene Name: | uncoupling protein 2 (mitochondrial, proton carrier) |
Gene Aliases: |
Slc25a8 |
Location: |
Chr.7:100493358-100499630 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 6 - Intron 6 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Ucp2 | NM_011671.5 | NP_035801.3 | ||
AF111998.1 | AAD17198.1 | |||
AF111999.1 | AAD17199.1 | |||
AK134116.1 | BAE22021.1 | |||
AK142687.1 | BAE25160.1 | |||
AK165245.1 | ||||
BC012697.1 | AAH12697.1 | |||
BC012967.1 | AAH12967.1 | |||
U69135.1 | AAB17666.1 | |||
U94593.1 | AAB53092.1 |
More Information
Set Membership: |
Intragenic Non-exonic |