Assay Details
Target Gene Details
Entrez Gene ID: | 12587 |
Gene Name: | melanoma inhibitory activity |
Gene Aliases: |
CD-RAP, Cdrap, Mia1 |
Location: |
Chr.7:27179741-27181149 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Mia | NM_019394.3 | 2 | 300 | NP_062267.2 |
AK135151.1 | 2 | 279 | BAE22443.1 | |
AW047492.1 | 3 | 284 | ||
BE864464.1 | 2 | 300 | ||
CK794645.1 | 2 | 294 | ||
X94322.1 | 2 | 305 | CAA63983.1 |