Assay Details
Target Gene Details
Entrez Gene ID: | 233877 |
Gene Name: | potassium channel tetramerisation domain containing 13 |
Gene Aliases: |
1500003N18Rik, AV259508, PDIP1alpha, Pdip1, Poldip1 |
Location: |
Chr.7:126928842-126947897 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Kctd13 | NM_172747.2 | NP_766335.1 | ||
AF534881.3 | AAO16177.1 | |||
AK075722.1 | BAC35910.1 | |||
BC079607.1 | AAH79607.1 |
More Information
Set Membership: |
Intragenic Non-exonic |