Assay Details
Target Gene Details
Entrez Gene ID: | 13107 |
Gene Name: | cytochrome P450, family 2, subfamily f, polypeptide 2 |
Gene Aliases: |
Cyp2f |
Location: |
Chr.7:27119955-27133660 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Cyp2f2 | NM_007817.2 | 3 | 336 | NP_031843.2 |
BC011089.1 | 3 | 307 | AAH11089.1 | |
BC024742.1 | 3 | 317 | AAH24742.1 | |
BU148924.1 | 3 | 300 | ||
M77497.1 | 3 | 316 | AAA37517.1 |
More Information
Set Membership: |
Intragenic Exonic |