Assay Details
Target Gene Details
Entrez Gene ID: | 233879 |
Gene Name: | aspartate beta-hydroxylase domain containing 1 |
Gene Aliases: |
A830007L07Rik, Gm168 |
Location: |
Chr.7:126945981-126949886 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Asphd1 | NM_001039645.1 | NP_001034734.1 | ||
AK043553.1 | ||||
AK134834.1 | ||||
BC111101.1 | AAI11102.1 |
Target Gene Details
Entrez Gene ID: | 233877 |
Gene Name: | potassium channel tetramerisation domain containing 13 |
Gene Aliases: |
1500003N18Rik, AV259508, PDIP1alpha, Pdip1, Poldip1 |
Location: |
Chr.7:126928842-126947897 on Build GRCm38 |
Assay Gene Location: |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Kctd13 |
More Information
Set Membership: |
Intragenic Non-exonic |