Assay Details
Target Gene Details
Entrez Gene ID: | 13088 |
Gene Name: | cytochrome P450, family 2, subfamily b, polypeptide 10 |
Gene Aliases: |
Cyp2b, Cyp2b20, p16 |
Location: |
Chr.7:25897658-25926624 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Cyp2b10 | NM_009999.4 | 1 | 102 | NP_034129.1 |
AF128849.1 | 1 | 84 | AAD28466.1 | |
AI304065.1 | 1 | 56 | ||
AK028103.1 | 1 | 102 | BAC25748.1 | |
BC060973.1 | 1 | 96 | AAH60973.1 | |
M21856.1 | 1 | 84 | AAA40425.1 | |
X99715.1 | 1 | 80 | CAA68051.1 |
More Information
Set Membership: |
Intragenic Exonic |