Assay Details
Target Gene Details
Entrez Gene ID: | 232414 |
Gene Name: | C-type lectin domain family 9, member a |
Gene Aliases: |
9830005G06Rik, DNGR-1 |
Location: |
Chr.6:129408862-129501701 on Build GRCm38 |
Assay Gene Location: |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Clec9a |
Target Gene Details
Entrez Gene ID: | 108078 |
Gene Name: | oxidized low density lipoprotein (lectin-like) receptor 1 |
Gene Aliases: |
LOX-1, SR-EI, Scare1 |
Location: |
Chr.6:129485247-129507165 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 6 - Exon 7 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Olr1 | NM_001301094.1 | NP_001288023.1 | ||
NM_001301096.1 | NP_001288025.1 | |||
NM_138648.2 | NP_619589.2 | |||
AF303744.1 | AAG44998.1 | |||
AK154687.1 | BAE32764.1 | |||
AK160919.1 | ||||
HQ603085.1 | ||||
HQ603086.1 |