Assay Details
Target Gene Details
Entrez Gene ID: | 27055 |
Gene Name: | FK506 binding protein 9 |
Gene Aliases: |
1810014L23, AW549641, FKBP-63, FKBP-9, FKBP60, FKBP63, FKBP65RS |
Location: |
Chr.6:56832059-56879360 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 2 - Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Fkbp9 | NM_012056.2 | NP_036186.2 | ||
AF090334.1 | AAC72964.1 | |||
AF279263.1 | AAF79215.1 | |||
AK184448.1 | ||||
AK198225.1 | ||||
AK198658.1 | ||||
AK204047.1 | ||||
AK206852.1 | ||||
AK208794.1 | ||||
AK209120.1 | ||||
AK209926.1 | ||||
AK214409.1 | ||||
AK216520.1 | ||||
BC026133.1 | AAH26133.1 | |||
BC043129.1 | AAH43129.1 |