Assay Details
Target Gene Details
Entrez Gene ID: | 69187 |
Gene Name: | endoplasmic reticulum protein 27 |
Gene Aliases: |
1810033M07Rik, 1810047B09Rik |
Location: |
Chr.6:136907314-136922461 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 5 - Exon 6 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Erp27 | NM_026983.2 | NP_081259.1 | ||
AK007684.1 | BAB25188.1 | |||
AK131830.1 | BAE20822.1 | |||
BC104406.1 | AAI04407.1 | |||
BC104407.2 | AAI04408.1 |
More Information
Set Membership: |
Intragenic Non-exonic |