Assay Details
Target Gene Details
Entrez Gene ID: | 56644 |
Gene Name: | C-type lectin domain family 7, member a |
Gene Aliases: |
BGR, Clecsf12, beta-GR |
Location: |
Chr.6:129461591-129472777 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Clec7a | NM_020008.2 | NP_064392.2 | ||
AA240643.1 | ||||
AF262985.1 | AAF72710.1 | |||
AY534909.1 | AAS37670.1 | |||
BC027742.1 | AAH27742.1 | |||
BY750022.1 |
Target Gene Details
Entrez Gene ID: | 232414 |
Gene Name: | C-type lectin domain family 9, member a |
Gene Aliases: |
9830005G06Rik, DNGR-1 |
Location: |
Chr.6:129408862-129501701 on Build GRCm38 |
Assay Gene Location: |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Clec9a |