Assay Details
Target Gene Details
Entrez Gene ID: | 71183 |
Gene Name: | C-type lectin domain family 12, member B |
Gene Aliases: |
4933425B16Rik |
Location: |
Chr.6:129375513-129427634 on Build GRCm38 |
Assay Gene Location: | Within Exon 6 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Clec12b | NM_001204223.1 | 6 | 724 | NP_001191152.1 |
NM_027709.2 | 5 | 638 | NP_081985.1 | |
AK016908.1 | 5 | 639 | BAB30491.1 | |
BC117712.2 | 6 | 684 |