Assay Details
Target Gene Details
Entrez Gene ID: | 231691 |
Gene Name: | serine dehydratase |
Gene Aliases: |
4432411H13Rik, SDH |
Location: |
Chr.5:120476547-120484935 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Sds | NM_145565.1 | 4 | 320 | NP_663540.1 |
AK149438.1 | 4 | 337 | BAE28875.1 | |
BC021605.1 | 4 | 317 | AAH21605.1 | |
BC021950.1 | 4 | 320 | AAH21950.1 |
More Information
Set Membership: |
Intragenic Exonic |