Assay Details
Target Gene Details
Entrez Gene ID: | 22256 |
Gene Name: | uracil DNA glycosylase |
Gene Aliases: |
UNG1, UNG2 |
Location: |
Chr.5:114130360-114139321 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Ung | NM_001040691.1 | NP_001035781.1 | ||
NM_011677.2 | NP_035807.2 | |||
AK146178.1 | BAE26956.1 | |||
AK168709.1 | BAE40551.1 | |||
AK168785.1 | BAE40619.1 | |||
BC011039.1 | AAH11039.1 | |||
BC052853.1 | AAH52853.1 | |||
U55041.1 | AAC53197.1 | |||
X99018.1 | CAA67489.1 | |||
Y08975.1 | CAA70168.1 |
More Information
Set Membership: |
Intragenic Non-exonic |