Assay Details
Target Gene Details
Entrez Gene ID: | 70103 |
Gene Name: | zinc finger, HIT domain containing 1 |
Gene Aliases: |
2700001K05Rik, ZNFN4A1 |
Location: |
Chr.5:136982194-136987882 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Znhit1 | NM_027318.3 | 2 | 486 | NP_081594.1 |
AK012199.1 | 2 | 126 | BAB28094.1 | |
AK080799.1 | 2 | 487 | BAC38025.1 | |
BC026751.1 | 2 | 132 | AAH26751.1 |
More Information
Set Membership: |
Intragenic Exonic |