Assay Details
Target Gene Details
Entrez Gene ID: | 26433 |
Gene Name: | procollagen-lysine, 2-oxoglutarate 5-dioxygenase 3 |
Gene Aliases: |
AI414586, LH3 |
Location: |
Chr.5:136987019-136996646 on Build GRCm38 |
Assay Gene Location: | Within Exon 11 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Plod3 | NM_011962.3 | 11 | 1851 | NP_036092.1 |
AF046783.1 | 11 | 1259 | AAD54618.1 | |
AK013195.1 | 11 | 1397 | BAB28704.1 | |
AK033360.1 | 11 | 1396 | BAC28246.1 | |
AK053320.1 | 11 | 1493 | ||
AK088948.1 | 11 | 1268 | BAC40669.1 | |
AK157741.1 | 10 | 1501 | ||
AK183180.1 | ||||
AK183516.1 | ||||
BC043047.1 | 11 | 1850 | AAH43047.1 | |
BC054734.1 | 11 | 1851 | AAH54734.1 |
More Information
Set Membership: |
Intragenic Exonic |