Assay Details
Target Gene Details
Entrez Gene ID: | 26394 |
Gene Name: | lysophospholipase 2 |
Gene Aliases: |
LysoII |
Location: |
Chr.4:135968224-135972594 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 6 - Exon 7 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Lypla2 | NM_011942.1 | NP_036072.1 | ||
AB009653.1 | BAA76751.1 | |||
AK003689.1 | BAB22940.1 | |||
AK075590.1 | BAC35841.1 | |||
AK089112.1 | BAC40757.1 | |||
AK167478.1 | BAE39559.1 | |||
BC068120.1 | AAH68120.1 |
More Information
Set Membership: |
Intragenic Non-exonic |