Assay Details
Target Gene Details
Entrez Gene ID: | 14748 |
Gene Name: | G-protein coupled receptor 3 |
Gene Aliases: |
Gpcr20, Gpcr21, Gpcr3 |
Location: |
Chr.4:133209340-133212536 on Build GRCm38 |
Assay Gene Location: | Overlaps - Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Gpr3 | NM_008154.3 | NP_032180.1 | ||
AK140302.1 | BAE24325.1 | |||
AK140464.1 | BAE24399.1 | |||
AK158917.1 | BAE34727.1 | |||
BB851645.1 |
More Information
Set Membership: |
Intragenic Non-exonic |