Assay Details
Target Gene Details
Entrez Gene ID: | 50798 |
Gene Name: | glucosamine (UDP-N-acetyl)-2-epimerase/N-acetylmannosamine kinase |
Gene Aliases: |
2310066H07Rik, DMRV, GLCNE, IBM2, NM, Uae1 |
Location: |
Chr.4:44034075-44084177 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Gne | NM_015828.3 | NP_056643.3 | ||
AK136573.1 | BAE23055.1 | |||
AM697710.1 | ||||
BC015277.1 | AAH15277.1 | |||
BC051254.1 | AAH51254.1 |
More Information
Set Membership: |
Intragenic Non-exonic |