Assay Details
Target Gene Details
Entrez Gene ID: | 100317 |
Gene Name: | expressed sequence AU040320 |
Gene Aliases: |
A730047D20Rik, Kiaa0319l |
Location: |
Chr.4:126753555-126869694 on Build GRCm38 |
Assay Gene Location: | Within Exon 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
AU040320 | NM_001035525.1 | 4 | 841 | NP_001030602.1 |
NM_001035526.1 | 4 | 959 | NP_001030603.1 | |
NM_133886.2 | 4 | 959 | NP_598647.1 | |
AK031342.1 | 4 | 903 | ||
AK043006.1 | 4 | 909 | BAC31432.1 | |
AK049570.1 | 4 | 842 | BAC33818.1 | |
AK084668.1 | 4 | 804 | BAC39244.1 | |
AK170261.1 | 4 | 916 | BAE41669.1 | |
BC028869.1 | 4 | 929 | AAH28869.1 | |
BM942797.1 | 3 | 614 | ||
CB249000.1 | 2 | 317 |