Assay Details
Target Gene Details
Entrez Gene ID: | 229905 |
Gene Name: | cysteine conjugate-beta lyase 2 |
Gene Aliases: |
KATIII, Kat3 |
Location: |
Chr.3:142701048-142744911 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 7 - Exon 8 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Ccbl2 | NM_001293560.1 | NP_001280489.1 | ||
NM_173763.4 | NP_776124.1 | |||
AF363737.1 | AAQ15190.1 | |||
AK049569.1 | BAC33817.1 | |||
AK145623.1 | BAE26546.1 | |||
BC131942.1 | ||||
BC132615.1 |
More Information
Set Membership: |
Intragenic Non-exonic |