Assay Details
Target Gene Details
Entrez Gene ID: | 78928 |
Gene Name: | phosphatidylinositol glycan anchor biosynthesis, class T |
Gene Aliases: |
2510012P17Rik, 4930534E15Rik, CGI-06, NDAP, Ndap7 |
Location: |
Chr.2:164497511-164508301 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Pigt | NM_133779.2 | 1 | 158 | NP_598540.2 |
AK019717.2 | 1 | 149 | ||
AK044499.1 | 1 | 142 | BAC31951.1 | |
AK049171.1 | 1 | 151 | ||
AK049990.1 | 1 | 151 | ||
AK090020.1 | 1 | 151 | ||
AK153462.1 | 1 | 144 | BAE32014.1 | |
AK153464.1 | 1 | 144 | BAE32015.1 | |
AK155057.1 | 1 | 145 | BAE33018.1 | |
AK157226.1 | 1 | 141 | BAE34008.1 | |
AK162573.1 | 1 | 138 | ||
BC034175.1 | 1 | 134 | AAH34175.1 | |
BU058874.1 | 1 | 158 |