Assay Details
Target Gene Details
Entrez Gene ID: | 16922 |
Gene Name: | phytanoyl-CoA hydroxylase |
Gene Aliases: |
AI256161, AI265699, Lnap1, PAHX |
Location: |
Chr.2:4918996-4938743 on Build GRCm38 |
Assay Gene Location: | Within Exon 6 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Phyh | NM_010726.2 | 6 | 625 | NP_034856.1 |
AF023463.1 | 6 | 590 | AAB81835.1 | |
AK146753.1 | 6 | 610 | BAE27410.1 | |
AK151372.1 | 6 | 573 | ||
AK164495.1 | 6 | 576 | BAE37810.1 | |
AK165412.1 | 6 | 569 | BAE38170.1 | |
AK168648.1 | 6 | 595 | BAE40505.1 | |
AK169213.1 | 6 | 567 | BAE40985.1 | |
BC002018.1 | 6 | 602 | AAH02018.1 | |
D88670.1 | 6 | 585 | BAA19003.1 |
More Information
Set Membership: |
Intragenic Exonic |