Assay Details
Target Gene Details
Entrez Gene ID: | 12716 |
Gene Name: | creatine kinase, mitochondrial 1, ubiquitous |
Gene Aliases: |
Mt-CK, ScCKmit, UbCKmit, mi-CK |
Location: |
Chr.2:121357751-121363737 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Ckmt1 | NM_009897.2 | 4 | 768 | NP_034027.1 |
AK005076.1 | 4 | 767 | BAB23800.1 | |
AK131953.1 | 4 | 772 | BAE20897.1 | |
BC025976.1 | 5 | 683 | AAH25976.1 | |
Z13968.1 | 4 | 768 | CAA78371.1 |
More Information
Set Membership: |
Intragenic Exonic |