Assay Details
Target Gene Details
Entrez Gene ID: | 65964 |
Gene Name: | sterile alpha motif and leucine zipper containing kinase AZK |
Gene Aliases: |
AV006891, B230120H23Rik, MLTK, MLTKalpha, MLTKbeta |
Location: |
Chr.2:72285637-72442610 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Zak | NM_001164791.1 | 3 | 398 | NP_001158263.1 |
NM_023057.5 | 3 | 462 | NP_075544.1 | |
NM_178084.4 | 3 | 398 | NP_835185.2 | |
AB049731.1 | 2 | 245 | BAB16442.1 | |
AB049732.1 | 2 | 245 | BAB16443.1 | |
AK045444.1 | 3 | 323 | BAC32371.1 | |
AK087363.1 | 3 | 337 | ||
AK132186.1 | 3 | 463 | BAE21021.1 | |
AK162819.1 | 3 | 263 | BAE37068.1 | |
AV470816.1 | 3 | 398 | ||
BC023718.1 | 3 | 343 | AAH23718.1 |
More Information
Set Membership: |
Intragenic Exonic |