Assay Details
Target Gene Details
Entrez Gene ID: | 50880 |
Gene Name: | selenocysteine lyase |
Gene Aliases: |
9830169H08, A930015N15Rik, SCL, Scly1, Scly2 |
Location: |
Chr.1:91298295-91321080 on Build GRCm38 |
Assay Gene Location: | Within Exon 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Scly | NM_016717.3 | 5 | 566 | NP_057926.2 |
AF175407.1 | 5 | 518 | AAF36812.1 | |
AK020864.1 | ||||
AK036533.1 | 5 | 535 | BAC29465.1 | |
AK036750.1 | 5 | 568 | BAC29563.1 | |
AK186846.1 | ||||
BC019879.1 | 5 | 526 | AAH19879.1 | |
BC021389.1 | 5 | 550 | AAH21389.1 |
More Information
Set Membership: |
Intragenic Exonic |