Assay Details
Target Gene Details
Entrez Gene ID: | 104009 |
Gene Name: | quiescin Q6 sulfhydryl oxidase 1 |
Gene Aliases: |
1300003H02Rik, QSOX, Qscn6, SOx |
Location: |
Chr.1:155778155-155812899 on Build GRCm38 |
Assay Gene Location: | Within Exon 6 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Qsox1 | NM_001024945.1 | 6 | 826 | NP_001020116.1 |
NM_023268.2 | 6 | 826 | NP_075757.1 | |
AB044284.1 | 6 | 734 | BAB21936.1 | |
AK004880.1 | 6 | 772 | BAB23638.1 | |
AK083938.1 | 6 | 777 | BAC39073.1 | |
AK166839.1 | 6 | 772 | BAE39061.1 | |
AK169877.1 | 6 | 779 | BAE41429.1 | |
AK169920.1 | 6 | 775 | BAE41459.1 | |
AK170449.1 | 6 | 739 | BAE41807.1 | |
AK204887.1 | 3 | 222 | ||
BC034131.1 | 3 | 318 | AAH34131.1 | |
BC076590.1 | 6 | 780 | AAH76590.1 |
More Information
Set Membership: |
Intragenic Exonic |