Assay Details
Target Gene Details
Entrez Gene ID: | 1621 |
Gene Name: | dopamine beta-hydroxylase |
Gene Aliases: |
DBM |
Location: |
Chr.9:133636363-133659344 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
DBH | NM_000787.3 | NP_000778.3 | ||
BC017174.2 | AAH17174.1 | |||
CD014125.1 | ||||
X13255.1 | CAA31631.1 | |||
X13256.1 | CAA31632.1 | |||
Y00096.1 | CAA68285.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv615639 | Chr.9:133639476 - 133654580 on Build GRCh38 | Loss | DBH |
esv2739138 | Chr.9:132727483 - 135521553 on Build GRCh38 | Deletion | DBH-AS1 FCN1 MRPS2 MIR3689B GTF3C5 MIR3689D2 MIR3689E CELP SARDH PPP1R26 SLC2A6 TSC1 ADAMTSL2 MED22 LOC105376306 RALGDS GBGT1 SPACA9 SURF4 FCN2 DBH COL5A1-AS1 TMEM8C SNORD36B OLFM1 C9orf62 OBP2B RNU6ATAC LOC401557 ABO PPP1R26-AS1 STKLD1 ADAMTS13 MIR3689C SNORD36C SURF2 MIR6877 CACFD1 SNORD141A SNORD36A COL5A1 LOC101928193 BRD3 LOC101448202 MIR3689F SNORD24 C9orf116 WDR5 RXRA SURF1 LINC00094 LOC101928525 MIR3689D1 VAV2 CEL AK8 GFI1B MIR3689A FAM163B MIR4669 MIR548AW LOC100996574 LCN1 RPL7A SURF6 LOC100130548 REXO4 |
nsv951800 | Chr.9:133627979 - 133696678 on Build GRCh38 | Deletion | DBH-AS1 SARDH DBH |
nsv469906 | Chr.9:133629444 - 133643961 on Build GRCh38 | Loss | DBH |
esv3576711 | Chr.9:133582804 - 133778331 on Build GRCh38 | Gain | DBH-AS1 VAV2 FAM163B SARDH DBH |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |